Anti-USP15 Goat Polyclonal Antibody

Supplier: Prosci
46-557
10116-840EA 701.94 USD
10116-840
Anti-USP15 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Immunohistochemistry: In paraffin embedded human liver shows patterned intracellular staining in hepatocytes. Recommended concentration, 2.5-3.8?g/ml

USP15 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: USP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR