Anti-NPRL2 Goat Polyclonal Antibody

Supplier: Prosci
46-539
10112-126EA 701.94 USD
10112-126
Anti-NPRL2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 45 kDa band observed in human muscle lysates (calculated MW of 43.7 kDa according to NP_006536.3). Recommended concentration :0. 1-0.3 ?g/mL.

TUSC4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TUSC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR