Anti-TRPC6 Goat Polyclonal Antibody

Supplier: Prosci
46-530
10112-990EA 701.94 USD
10112-990
Anti-TRPC6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 80 kDa band in human spleen and testis lysates after 1 ?g/mL antibody staining.2. The 80 kDa band was successfully blocked by incubation with the immunizing peptide.

TRPC6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TRPC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR