Anti-TBPL1 Goat Polyclonal Antibody

Supplier: Prosci
46-514
10112-970EA 701.94 USD
10112-970
Anti-TBPL1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Approximately 20 kDa band seen in Jurkat lysate [Predicted MW of 20 kDa according to NP_004856]. Recommended for use at 1-3 ?g/mL.

TRF2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TBPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR