Anti-TIRAP Goat Polyclonal Antibody

Supplier: Prosci
46-500
10112-122EA 701.94 USD
10112-122
Anti-TIRAP Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 30 kDa band observed in human liver lysates (calculated MW of 25.3 kDa according to NP_683708.1). Recommended concentration: 1-3 ?g/mL.

TIRAP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TIRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR