Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRIM24 Goat Polyclonal Antibody

Supplier: Prosci
46-493
10112-934
Anti-TRIM24 Goat Polyclonal Antibody
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 120 kDa band in human kidney and heart lysates at 1 μg/mL, however prominent bands were also seen at 25 kDa and 45 kDa . Immunohistochemistry: In paraffin embedded human breast carcinoma shows nuclear staining in the distorted lobules. Recommended concentration, 4-6μg/ml

TIF1 alpha antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TIF1 alpH a antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRIM24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty