Anti-TFE3 Goat Polyclonal Antibody

Supplier: Prosci
46-482
10112-918EA 701.94 USD
10112-918
Anti-TFE3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Approximately 60 kDa band observed in lysates of HEK293 (calculated MW of 61.5 kDa according to NP_006512.2). Recommended concentration: 1-3 ?g/mL.

TFE3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TFE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR