Anti-TNS1 Goat Polyclonal Antibody

Supplier: Prosci
46-479
10112-912EA 701.94 USD
10112-912
Anti-TNS1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2,000. Immunohistochemistry: In paraffin embedded human kidney shows weak staining of the basement membranes of convoluted tubules. Recommended concentration, 4-6?g/ml

Tensin-1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR