Anti-TAGLN Goat Polyclonal Antibody

Supplier: Prosci
46-467
10112-894EA 701.94 USD
10112-894
Anti-TAGLN Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 23 kDa band observed in human heart, placenta and testis lysates (calculated MW of 22.6 kDa according to NP_003177.2 and NP_001001522.1). Recommended concentration: 0.001-0.003 ?g/mL.

TAGLN antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TAGLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR