Anti-SDCBP Goat Polyclonal Antibody

Supplier: Prosci
46-459
10112-880EA 701.94 USD
10112-880
Anti-SDCBP Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 28-30 kDa band observed in lysates of cell lines HEK293 and A431 (calculated MW of 32.4 kDa according to NP_005616.2 and NP_001007068.1 and of 32.3 kDa according to NP_001007070.1 and NP_001007071.1). Recommended concentration: 0.01-0.03 ?g/mL.

Syntenin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SDCBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR