Anti-UBASH3A Goat Polyclonal Antibody

Supplier: Prosci
46-446
10112-858EA 701.94 USD
10112-858
Anti-UBASH3A Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 48 kDa band in lysates of B-Daudi lysate after 0.1 ?g/mL antibody staining.1 kDa according to NP_061834.1 and 69.8 kDa according to NP_001001895.1. The 48 kDa band was successfully blocked by incubation with the immunizing peptide.

STS2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: UBASH3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR