Anti-STELLAR Goat Polyclonal Antibody

Supplier: Prosci
46-444
10116-822EA 552.17 USD
10116-822
Anti-STELLAR Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 26 kDa band in human ovary and placenta lysates after 0.1 ?g/mL antibody staining. The 26 kDa band was successfully blocked by incubation with the immunizing peptide.

STELLAR antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STELLAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR