Anti-STAP2 Goat Polyclonal Antibody

Supplier: Prosci
46-439
10116-814EA 701.94 USD
10116-814
Anti-STAP2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 48 kDa band observed in human heart lysates (calculated MW of 49.6 kDa according to NP_060190.2). Recommended concentration: 0.05-0.3 ?g/mL. Immunohistochemistry: In paraffin embedded human cortex shows textured cytoplasm staining in neuronal bodies and processes. Recommended concentration, 1-2?g/ml

STAP-2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR