Anti-STAG2 Goat Polyclonal Antibody

Supplier: Prosci
46-436
10116-810EA 701.94 USD
10116-810
Anti-STAG2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 150 kDa band observed in HeLa lysate (calculated MW of 146 kDa according to NP_001036214.1 and NP_001036215.1). Recommended concentration: 0.01 ?g/mL.

STAG2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STAG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR