Anti-SSX2 Goat Polyclonal Antibody

46-435
10116-808EA 689.45 USD
10116-808
Anti-SSX2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. this antibody has successfully been used on cell line K562 in immunocytochemistry/Immunofluorescence. See Denniss et al, Br J Haematol. 2007 Sep;138 (5) :668-9. (PMID: 17686061). In Western Blot we find no specific signal but low background (at antibody concentration up to 1 ?g/mL) in K562 lysate .

SSX2IP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SSX2IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR