Anti-SORD Goat Polyclonal Antibody

Supplier: Prosci
46-413
10116-782EA 701.94 USD
10116-782
Anti-SORD Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 38 kDa band observed in mouse kidney lysates (calculated MW of 38.3 kDa according to NP_003095.1). Recommended concentration: 0.1-1 ?g/mL.

Sorbitol Dehydrogenase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SORD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR