Anti-SNX16 Goat Polyclonal Antibody

Supplier: Prosci
46-399
10116-764EA 701.94 USD
10116-764
Anti-SNX16 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 38 kDa band observed in lysates of Daudi lysate (calculated MW of 39.2 kDa according to NP_071416.2). In transfected HEK293 transiently expressing SNX16 a band of approximately 40 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.05-0.2 ?g/mL.

SNX16 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SNX16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR