Anti-SNX15 Goat Polyclonal Antibody

Supplier: Prosci
46-398
10116-762EA 701.94 USD
10116-762
Anti-SNX15 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 48 kDa band seen in human kidney lysates and in HEK293 lysate (calculated MW 38.2 kDa according to NP_037438.2). Recommended for use at 0.2-1 ?g/mL. Note: A single Approximately 30-35 kDa band (probably representing the shorter isoform) was observed in human heart.

SNX15 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SNX15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR