Anti-SNX12 Goat Polyclonal Antibody

Supplier: Prosci
46-397
10116-760EA 701.94 USD
10116-760
Anti-SNX12 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution >1:32,000. Immunohistochemistry: In paraffin embedded human breast shows vesiculate staining in the cytoplasm of epithelial cells in one lobule. Similar results were obtained in human Liver. Recommended concentration, 3-6 ?g/mL.

SNX12 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SNX12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR