Anti-SNAI1 Goat Polyclonal Antibody

Supplier: Prosci
46-392
10116-756EA 701.94 USD
10116-756
Anti-SNAI1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Approximately 28 kDa band observed in lysates A431 and human kidney lysate (calculated MW of 29.1 kDa according to NP_005976.2). Recommended concentration: 0.1-0.3 ?g/mL. This antibody was successfully used in immunohistochemistry on mouse lung adenocarcinoma induced by urethane.

SNAI1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SNAI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR