Anti-HTR3A Goat Polyclonal Antibody

Supplier: Prosci
46-357
10116-700EA 701.94 USD
10116-700
Anti-HTR3A Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 55 kDa band observed in human colon lysates (calculated MW of 55.3 kDa according to NP_000860.1. Recommended concentration: 0.3-1 ?g/mL. A minor band of unknown identity was also consistently observed at 200 kDa . This band was successfully blocked by incubation with the immunizing peptide.

Serotonin receptor 3A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HTR3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR