Anti-SART3 Goat Polyclonal Antibody

Supplier: Prosci
46-338
10116-666EA 701.94 USD
10116-666
Anti-SART3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Approximately 170 kDa band observed in lysates of cell lines Daudi, Jurkat and MOLT4 (calculated MW of 111 kDa according to NP_055521.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Yang et al, tumor Res. 1999 Aug 15;59 (16) :4056-63; PMID: 10463607). Recommended concentration: 0.01-0.03 ?g/mL.

SART3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SART3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR