Anti-SALL4 Goat Polyclonal Antibody

Supplier: Prosci
46-333
10116-656EA 701.94 USD
10116-656
Anti-SALL4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 70 kDa band observed in rat spleen and testis lysates (calculated MW of 66.2 kDa according to NP_958797.2). Recommended concentration: 0.1-0.3 ?g/mL. Additional bands of unknown identity was also consistently observed at 28+24 kDa . These bands were successfully blocked by incubation with the immunizing peptide.

SALL4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Sall4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR