Anti-SAE1 Goat Polyclonal Antibody

Supplier: Prosci
46-328
10116-648EA 701.94 USD
10116-648
Anti-SAE1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 40 kDa band observed in Jurkat and 293 cell lysates (predicted MW of 40 kDa according to NP_005491). Recommended for use at 1 ?g/mL.

SAE1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SAE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR