Anti-KIAA1881 Goat Polyclonal Antibody

Supplier: Prosci
46-327
10116-646EA 701.94 USD
10116-646
Anti-KIAA1881 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 45 kDa and 50 kDa in human adipose lysates after 0.1 ?g/mL antibody staining.1. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands). Immunohistochemistry: In paraffin embedded human pancreas shows membranous and cytoplasm staining in adipocytes. Recommended concentration, 3-6 ?g/mL.

S3-12 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KIAA1881
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR