Anti-S100 A4 Goat Polyclonal Antibody

Supplier: Prosci
46-326
10116-644EA 701.94 USD
10116-644
Anti-S100 A4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 13 kDa band observed in human placenta lysates (calculated MW of 11.7 kDa according to NP_002952.1and NP_062427.1). Recommended concentration: 0.03-0.1 ?g/mL.

S100A4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: S100A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR