Anti-RPS19 Goat Polyclonal Antibody

Supplier: Prosci
46-322
10116-638EA 701.94 USD
10116-638
Anti-RPS19 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 15 kDa band observed in human lysates of cell lines Daudi and K562 (calculated MW of 16.1 kDa according to NP_001013.1). In transfected HEK293 transiently expressing RPS19 a band of approximately 18 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.03-0.1 ?g/mL.

RPS19 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RPS19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR