Anti-RPE65 Goat Polyclonal Antibody

Supplier: Prosci
46-321
10116-636EA 701.94 USD
10116-636
Anti-RPE65 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 60 kDa band observed in rat retina lysates (calculated MW of 60.9 kDa according to human NP_000320.1 and 61.0 kDa according to rat NP_446014.1). Recommended concentration: 0.5-1.5 ?g/mL.

RPE65 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RPE65
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR