Anti-RPL19 Goat Polyclonal Antibody

46-295
10116-602EA 689.45 USD
10116-602
Anti-RPL19 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 25 kDa band observed in human testis lysates (calculated MW of 23.5 kDa according to NP_000972.1). Recommended concentration: 0.1-0.3 ?g/mL.

Ribosomal Protein L19 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RPL19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR