Anti-RGS1 Goat Polyclonal Antibody

Supplier: Prosci
46-287
10116-592EA 701.94 USD
10116-592
Anti-RGS1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 25 kDa band seen in HepG2 lysate. Recommended for use at 0.5-2 ?g/mL.

RGS1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RGS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR