Anti-RNLS Goat Polyclonal Antibody

Supplier: Prosci
46-278
10112-070EA 701.94 USD
10112-070
Anti-RNLS Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 37 kDa band observed in human kidney and mouse kidney lysates (calculated MW of 37.8 kDa according to NP_001026879.1 and 34.9 kDa according to NP_060833.1). Recommended concentration: 0.1-0.3 ?g/mL.

Renalase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RNLS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR