Anti-RAD51L3 Goat Polyclonal Antibody

Supplier: Prosci
46-261
10116-562EA 552.17 USD
10116-562
Anti-RAD51L3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 65 kDa band in human brain and A431 lysates at 1 ?g/mL, this band was successfully blocked by incubation with the immunizing peptide.

RAD51D antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RAD51L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR