Anti-PC Goat Polyclonal Antibody

Supplier: Prosci
46-255
10116-550EA 701.94 USD
10116-550
Anti-PC Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 150 kDa band observed in mouse liver lysates (calculated MW of 130 kDa according to NP_000911.2, NP_001035806.1and NP_071504.2). Recommended concentration: 0.03-0.1 ?g/mL.

Pyruvate carboxylase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR