Anti-S100 A7 Goat Polyclonal Antibody

46-241
10116-534EA 689.45 USD
10116-534
Anti-S100 A7 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 15 kDa band in human skin lysates after 0.3 ?g/mL antibody staining.5 kDa according to NP_002954.1. The 15 kDa band was successfully blocked by incubation with the immunizing peptide.

Psoriasin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: S100A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR