Anti-TTR Goat Polyclonal Antibody

Supplier: Prosci
46-219
10116-508EA 701.94 USD
10116-508
Anti-TTR Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 20-22 kDa band in human heart, kidney, liver, lung, placenta and testis lysates after 1 ?g/mL antibody staining.The 20-22 kDa band was successfully blocked by incubation with the immunizing peptide.

Prealbumin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TTR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR