Anti-PPP2R5E Goat Polyclonal Antibody

Supplier: Prosci
46-212
10116-496EA 701.94 USD
10116-496
Anti-PPP2R5E Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 55 kDa and 40 kDa bands seen in mouse brain extracts. Recommended for use at 0.5-2 ?g/mL. Please note that the two bands may correspond to two different isoforms. Two isoforms of the epsilon mRNA have been detected in Northern blots (J Biol Chem 1996 Sep 6;271 (36) :22081-9).

PPP2R5E antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PPP2R5E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR