Anti-PLEK Goat Polyclonal Antibody

Supplier: Prosci
46-201
10116-474EA 701.94 USD
10116-474
Anti-PLEK Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 45 kDa band observed in human heart and PBMC lysates (predicted MW of 40 kDa according to NP_002655). Recommended for use at 0.05-1 ?g/mL.

Pleckstrin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PLEK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR