Anti-PINK1 Goat Polyclonal Antibody

Supplier: Prosci
46-190
10116-456EA 701.94 USD
10116-456
Anti-PINK1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 55 kDa band observed in rat testis lysates (calculated MW of 62.8 kDa according to NP_115785.1). Recommended concentration: 1-3 ?g/mL.

PINK1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PINK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR