Anti-PLIN1 Goat Polyclonal Antibody

Supplier: Prosci
46-167
10112-046EA 701.94 USD
10112-046
Anti-PLIN1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 60 kDa band observed in human adipose lysates (calculated MW of 56.0 kDa according to NP_002657.2). Recommended concentration: 0.01 ?g/mL.

Perilipin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PLIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR