Anti-PAX8 Goat Polyclonal Antibody

Supplier: Prosci
46-141
10112-034EA 701.94 USD
10112-034
Anti-PAX8 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: an approximately 40 kDa band observed in Human Liver lysates. Recommended concentration: 1-3 ?g/mL. Immunohistochemistry: In paraffin embedded Human Thyroid Gland shows nuclear staining in epithelial cells around follicles. Recommended concentration, 1-2 ?g/mL. Does not stain mouse tissues

PAX8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PAX8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR