Anti-PLDN Goat Polyclonal Antibody

Supplier: Prosci
46-128
10112-828EA 701.94 USD
10112-828
Anti-PLDN Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 23-24 kDa band observed in lysates of cell line A431 (calculated MW of 19.7 kDa according to NP_036520.1). In transfected HEK293 transiently expressing PLDN a band of approximately 28 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 1-3 ?g/mL.

Pallidin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PLDN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR