Anti-ORC6L Goat Polyclonal Antibody

Supplier: Prosci
46-103
10112-786EA 701.94 USD
10112-786
Anti-ORC6L Goat Polyclonal Antibody
Antibodies
Western Blot: Approximately 30-35 kDa band seen in HeLa cell lysates. Recommended for use at 0.2-1 ?g/mL.

ORC6L antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ORC6L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR