Anti-ORC4 Goat Polyclonal Antibody

Supplier: Prosci
46-102
10112-784EA 701.94 USD
10112-784
Anti-ORC4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 40-45 kDa band seen in Jurkat and NSO (mouse) cell lysates. Recommended for use at 1-3 ?g/mL.

ORC4L antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ORC4L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR