Anti-ORC3 Goat Polyclonal Antibody

Supplier: Prosci
46-101
10112-782EA 701.76 USD
10112-782
Anti-ORC3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 75 kDa band observed in lysates of cell lines HepG2 and HeLa (calculated MW of 82.3 kDa according to NP_862820.1 and NP_036513.2). Recommended concentration: 1-3 ?g/mL. an additional band of unknown identity was also consistently observed at 35 kDa . This band was successfully blocked by incubation with the immunizing peptide.

ORC3L antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ORC3L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR