Anti-NUP62 Goat Polyclonal Antibody

Supplier: Prosci
45-986
10112-756EA 552.17 USD
10112-756
Anti-NUP62 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 30 kDa band in human testis lysates after 0.5 ?g/mL antibody staining. The 30 kDa band was successfully blocked by incubation with the immunizing peptide.

NUP62 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NUP62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR