Anti-NUP50 Goat Polyclonal Antibody

Supplier: Prosci
45-985
10112-754EA 701.76 USD
10112-754
Anti-NUP50 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 60 kDa band observed in Jurkat lysate (calculated MW 50.1 kDa according to NP_009103.2). Recommended for use at 0.2-1 ?g/mL. Immunohistochemistry: In paraffin embedded human uterus shows textured nuclear staining of endometrial cells. Recommended concentration, 2.5-3.8 ?g/mL.

Nup50 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NUP50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR