Anti-CARD15 Goat Polyclonal Antibody

Supplier: Prosci
45-973
10112-734EA 701.94 USD
10112-734
Anti-CARD15 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 100 kDa band observed in lysates of human peripheral blood mononucleocytes (calculated MW of 115 kDa according to NP_071445.1). Recommended concentration: 0.1-0.3 ?g/mL. an additional band of unknown identity was also consistently observed at 60 kDa . This band was successfully blocked by incubation with the immunizing peptide.

NOD2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CARD15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR