Anti-NFIL3 Goat Polyclonal Antibody

Supplier: Prosci
45-965
10112-724EA 701.94 USD
10112-724
Anti-NFIL3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 55-60 kDa band observed in lysates of Jurkat and HepG2 (calculated MW of 51.5 kDa according to NP_005375.2). The observed molecular weight corresponds to earlier findings in literature (Ikushima et al., Proc Natl Acad Sci U S A. 1997 Mar 18;94 (6) :2609-14.; PMID: 9122243). Recommended concentration: 0.5-1.5 ?g/mL.

NFIL3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NFIL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR