Anti-SERPINI1 Goat Polyclonal Antibody

Supplier: Prosci
45-961
10112-720EA 701.94 USD
10112-720
Anti-SERPINI1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 48 kDa band observed in human brain lysate (calculated MW of 46.4 kDa according to NP_005016.1). Recommended concentration: 1-3 ?g/mL.

Neuroserpin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SERPINI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR