Anti-NPY5R Goat Polyclonal Antibody

Supplier: Prosci
45-960
10112-718EA 701.94 USD
10112-718
Anti-NPY5R Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 50 kDa band observed in human brain, breast and duodenum lysates (calculated MW of 50.7 kDa according to NP_006165.1). Recommended concentration: 0.3-1 ?g/mL. Immunohistochemistry: In paraffin embedded Human Intestine shows staining of ganglion cells. Recommended concentration, 4-6 ?g/mL.

Neuropeptide Y Receptor Y5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NPY5R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR